Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wfdc6a
Entrez Gene ID
685153
Gene Name
WAP four-disulfide core domain 6A
Gene Aliases
-
Chromosome Location
Chr.3: 161036703 - 161040511 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA GQ406387.1 - 1 - 2 128 98

View Full Product Details