Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cpsf7
Entrez Gene ID
365407
Gene Name
cleavage and polyadenylation specific factor 7
Gene Aliases
RGD1305441
Chromosome Location
Chr.1: 226572389 - 226596402 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014245.2 NP_001014267.1 4 - 5 579 96
XM_008760239.1 XP_008758461.1 5 - 6 653 96
GenBank mRNA BC083864.1 - 4 - 5 582 96

View Full Product Details