The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001104560.2 | NP_001098030.1 | 3 - 4 | 699 | 74 |
GenBank mRNA | AB297662.1 | - | 3 - 4 | 700 | 74 |