Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wnt3a
Entrez Gene ID
303181
Gene Name
wingless-type MMTV integration site family, member 3A
Gene Aliases
-
Chromosome Location
Chr.10: 45598898 - 45643151 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107005.2 NP_001100475.2 1 - 2 87 96

View Full Product Details