Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Triml2
Entrez Gene ID
290766
Gene Name
tripartite motif family-like 2
Gene Aliases
RGD1565382
Chromosome Location
Chr.16: 52116975 - 52127591 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191690.1 NP_001178619.1 4 - 5 619 97

View Full Product Details