Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tsga10ip
Entrez Gene ID
361707
Gene Name
testis specific 10 interacting protein
Gene Aliases
-
Chromosome Location
Chr.1: 220772592 - 220786931 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270736.1 NP_001257665.1 7 - 8 1473 58

View Full Product Details