Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Frmd6
Entrez Gene ID
257646
Gene Name
FERM domain containing 6
Gene Aliases
-
Chromosome Location
Chr.6: 93281382 - 93358888 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271054.1 NP_001257983.1 7 - 8 946 79
XM_006240151.2 XP_006240213.1 7 - 8 996 79
XM_006240152.2 XP_006240214.1 8 - 9 1076 79
XM_006240153.2 XP_006240215.1 7 - 8 1420 79
GenBank mRNA AF441249.1 - 6 - 7 723 79

View Full Product Details