Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pstk
Entrez Gene ID
361661
Gene Name
phosphoseryl-tRNA kinase
Gene Aliases
RGD1564300
Chromosome Location
Chr.1: 201950148 - 201960563 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271229.1 NP_001258158.1 4 - 5 818 115
GenBank mRNA BC168162.1 - 3 - 4 1052 115

View Full Product Details