Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cyp2c24
Entrez Gene ID
499353
Gene Name
cytochrome P450, family 2, subfamily c, polypeptide 24
Gene Aliases
RGD1563697
Chromosome Location
Chr.1: 258074860 - 258139176 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271354.1 NP_001258283.1 2 - 3 353 106

View Full Product Details