Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Psma6
Entrez Gene ID
29673
Gene Name
proteasome (prosome, macropain) subunit, alpha type 6
Gene Aliases
-
Chromosome Location
Chr.6: 76174462 - 76205429 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017283.3 NP_058979.1 5 - 6 643 98
XM_006240098.2 XP_006240160.1 5 - 6 601 98
GenBank mRNA BC062232.1 - 5 - 6 644 98
D10755.1 - 5 - 6 632 98
FQ212808.1 - - 745 98
FQ220193.1 - 5 - 6 670 98

View Full Product Details