Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ndufv2
Entrez Gene ID
81728
Gene Name
NADH dehydrogenase (ubiquinone) flavoprotein 2
Gene Aliases
-
Chromosome Location
Chr.9: 113875718 - 113900169 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031064.2 NP_112326.1 5 - 6 520 60
GenBank mRNA BC058495.1 - 5 - 6 524 60
M22756.1 - 5 - 6 445 60

View Full Product Details