Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Star
Entrez Gene ID
25557
Gene Name
steroidogenic acute regulatory protein
Gene Aliases
-
Chromosome Location
Chr.16: 71036204 - 71040847 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031558.3 NP_113746.1 1 - 2 186 74
GenBank mRNA AB001349.1 - - 142 74
AB006007.1 - 1 - 2 187 74
AF044081.1 - - 126 74
AY736357.1 - 1 - 2 67 74
BC088859.1 - 1 - 2 78 74

View Full Product Details