Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Xbp1
Entrez Gene ID
289754
Gene Name
X-box binding protein 1
Gene Aliases
HTF
Chromosome Location
Chr.14: 85753736 - 85758820 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004210.2 NP_001004210.1 4 - 4 539 76
GenBank mRNA BC079450.1 - 4 - 4 520 76
BU670819.1 - 4 - 4 539 76

View Full Product Details