Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Crebzf
- Entrez Gene ID
-
293112
- Gene Name
- CREB/ATF bZIP transcription factor
- Gene Aliases
-
RGD1565584
- Chromosome Location
-
Chr.1: 156290215 - 156296891 on Build Rnor_6.0
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
CB700395.1 |
- |
|
185 |
|
Chr. - - [-] on Build |
90 |
GenBankmRNA |
BG669877.1 |
- |
|
227 |
|
Chr. - - [-] on Build |
90 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
transmembrane protein 126B |
Tmem126b |
RGD1308371 |
-16.0kb
|
Anti-sense |
Chr. - 156262220 - 156274207 on Build Rnor_6.0 |
Downstream |
coiled-coil domain-containing protein 89-like |
LOC100911776 |
|
31.6kb
|
Sense |
Chr. - 156325241 - 156328892 on Build Rnor_6.0 |
View Full Product Details