Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pex13
Entrez Gene ID
305581
Gene Name
peroxisomal biogenesis factor 13
Gene Aliases
-
Chromosome Location
Chr.14: 108394299 - 108411994 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107242.2 NP_001100712.1 2 - 3 833 86
GenBank mRNA FM088835.1 - 1 - 2 482 86

View Full Product Details