Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nppa
Entrez Gene ID
24602
Gene Name
natriuretic peptide A
Gene Aliases
ANF, ANP, Pnd, RATANF
Chromosome Location
Chr.5: 164808407 - 164809716 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012612.2 NP_036744.1 3 - 3 724 108
GenBank mRNA AI602287.1 - 1 - 1 108
BC158590.1 - 3 - 3 714 108
M15868.1 - 2 - 2 538 108
M27498.1 - 3 - 3 712 108
X00665.1 - 3 - 3 715 108
X01118.1 - 3 - 3 704 109

View Full Product Details