The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031116.3 | NP_112378.2 | 2 - 3 | 237 | 74 |
GenBank mRNA | BC058138.1 | - | 2 - 3 | 253 | 74 |
EF121972.1 | - | - | 195 | 74 | |
EF122007.1 | - | 2 - 3 | 195 | 74 | |
U06436.1 | - | 2 - 3 | 195 | 74 |