Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MDM10
Entrez Gene ID
851223
Gene Name
Gene Aliases
FUN37, YAL010C
Chromosome Location
Chr.I: 134184 - 135665 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178155.2 NP_009392.2 1 - 1 1348 108

View Full Product Details