Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ACS1
Entrez Gene ID
851245
Gene Name
Gene Aliases
FUN44, YAL054C
Chromosome Location
Chr.I: 42881 - 45022 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178197.1 NP_009347.1 1 - 1 2050 77

View Full Product Details