Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BDH2
Entrez Gene ID
851238
Gene Name
Gene Aliases
YAL061W
Chromosome Location
Chr.I: 33448 - 34701 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178203.1 NP_009340.1 1 - 1 1152 75

View Full Product Details