Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SSA3
Entrez Gene ID
852203
Gene Name
Gene Aliases
YBL075C, YG106
Chromosome Location
Chr.II: 84499 - 86448 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178315.1 NP_009478.1 1 - 1 1851 103

View Full Product Details