Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TRM7
Entrez Gene ID
852353
Gene Name
Gene Aliases
YBR061C
Chromosome Location
Chr.II: 364787 - 365719 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178409.3 NP_009617.3 1 - 1 844 56

View Full Product Details