Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VID24
Entrez Gene ID
852402
Gene Name
Gene Aliases
GID4, YBR105C
Chromosome Location
Chr.II: 450881 - 451969 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178453.1 NP_009663.1 1 - 1 948 70

View Full Product Details