Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TFC1
Entrez Gene ID
852421
Gene Name
Gene Aliases
YBR123C
Chromosome Location
Chr.II: 484742 - 486691 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178471.3 NP_009681.3 1 - 1 1846 116

View Full Product Details