Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VMA2
Entrez Gene ID
852424
Gene Name
Gene Aliases
VAT2, YBR127C
Chromosome Location
Chr.II: 491269 - 492822 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178475.3 NP_009685.3 1 - 1 1252 71

View Full Product Details