Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KTR4
Entrez Gene ID
852498
Gene Name
Gene Aliases
YBR199W
Chromosome Location
Chr.II: 618909 - 620303 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178547.3 NP_009758.3 1 - 1 1249 103

View Full Product Details