Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AME1
Entrez Gene ID
852512
Gene Name
Gene Aliases
ARP100, YBR211C
Chromosome Location
Chr.II: 646158 - 647132 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178559.3 NP_009770.3 1 - 1 847 100

View Full Product Details