Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HPC2
Entrez Gene ID
852516
Gene Name
Gene Aliases
YBR215W
Chromosome Location
Chr.II: 653356 - 655317 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178563.3 NP_009774.4 1 - 1 1748 69

View Full Product Details