Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RGD1
Entrez Gene ID
852563
Gene Name
Gene Aliases
YBR260C
Chromosome Location
Chr.II: 732639 - 734639 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178608.1 NP_009819.1 1 - 1 1950 85

View Full Product Details