Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TSC10
Entrez Gene ID
852568
Gene Name
Gene Aliases
YBR265W
Chromosome Location
Chr.II: 738582 - 739544 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178613.2 NP_009824.2 1 - 1 650 96

View Full Product Details