Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HSM3
Entrez Gene ID
852575
Gene Name
Gene Aliases
YBR272C
Chromosome Location
Chr.II: 746361 - 747803 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178620.3 NP_009831.3 1 - 1 1345 86

View Full Product Details