Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
APE3
Entrez Gene ID
852589
Gene Name
Gene Aliases
APY1, YBR286W
Chromosome Location
Chr.II: 774700 - 776313 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178634.1 NP_009845.2 1 - 1 1549 91

View Full Product Details