Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IMG1
Entrez Gene ID
850413
Gene Name
Gene Aliases
YCR046C, bL19m
Chromosome Location
Chr.III: 209914 - 210423 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178760.1 NP_009975.1 1 - 1 248 62

View Full Product Details