Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HCM1
Entrez Gene ID
850429
Gene Name
Gene Aliases
YCR065W
Chromosome Location
Chr.III: 229310 - 231004 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178776.1 NP_009991.2 1 - 1 1553 86

View Full Product Details