Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SSK22
Entrez Gene ID
850436
Gene Name
Gene Aliases
YCR073C
Chromosome Location
Chr.III: 242588 - 246583 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178783.1 NP_009998.2 1 - 1 3850 70

View Full Product Details