Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CDC50
Entrez Gene ID
850456
Gene Name
Gene Aliases
YCR094W
Chromosome Location
Chr.III: 286762 - 287937 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178800.1 NP_010018.1 1 - 1 551 73

View Full Product Details