Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
YET3
Entrez Gene ID
851487
Gene Name
Gene Aliases
YDL072C
Chromosome Location
Chr.IV: 329836 - 330447 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180131.1 NP_010211.1 1 - 1 543 85

View Full Product Details