Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MRK1
Entrez Gene ID
851480
Gene Name
Gene Aliases
YDL079C
Chromosome Location
Chr.IV: 312951 - 314748 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180138.1 NP_010204.1 1 - 2 407 117

View Full Product Details