Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATG20
Entrez Gene ID
851445
Gene Name
Gene Aliases
CVT20, SNX42, YDL113C
Chromosome Location
Chr.IV: 256633 - 258555 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180172.1 NP_010170.1 1 - 1 1642 98

View Full Product Details