Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IWR1
Entrez Gene ID
851442
Gene Name
Gene Aliases
YDL115C
Chromosome Location
Chr.IV: 253995 - 255126 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180174.1 NP_010168.4 1 - 2 86 139

View Full Product Details