Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ASF2
Entrez Gene ID
851330
Gene Name
Gene Aliases
YDL197C
Chromosome Location
Chr.IV: 104917 - 106494 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180257.1 NP_010084.1 1 - 1 1448 70

View Full Product Details