Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NOP6
Entrez Gene ID
851313
Gene Name
Gene Aliases
YDL213C
Chromosome Location
Chr.IV: 77289 - 77966 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180273.1 NP_010068.1 1 - 1 450 83

View Full Product Details