Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PTP1
Entrez Gene ID
851368
Gene Name
Gene Aliases
YDL230W
Chromosome Location
Chr.IV: 42700 - 43707 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180290.1 NP_010051.1 1 - 1 943 76

View Full Product Details