Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPO71
Entrez Gene ID
851681
Gene Name
Gene Aliases
YDR104C
Chromosome Location
Chr.IV: 661173 - 664910 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180412.3 NP_010389.3 1 - 1 3644 97

View Full Product Details