Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KEX1
Entrez Gene ID
852670
Gene Name
Gene Aliases
YGL203C
Chromosome Location
Chr.VII: 112475 - 114664 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181068.1 NP_011312.1 1 - 1 2046 78

View Full Product Details