Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DPB4
Entrez Gene ID
851699
Gene Name
Gene Aliases
YDR121W
Chromosome Location
Chr.IV: 693585 - 694175 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180429.3 NP_010406.3 1 - 1 450 99

View Full Product Details