Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
YCF1
Entrez Gene ID
851713
Gene Name
Gene Aliases
YDR135C
Chromosome Location
Chr.IV: 723004 - 727551 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180442.3 NP_010419.3 1 - 1 4449 86

View Full Product Details