Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NUM1
Entrez Gene ID
851727
Gene Name
Gene Aliases
PAC12, YDR150W
Chromosome Location
Chr.IV: 755628 - 763874 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180457.1 NP_010434.1 1 - 1 8149 59

View Full Product Details