Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SEC5
Entrez Gene ID
851744
Gene Name
Gene Aliases
YDR166C
Chromosome Location
Chr.IV: 786306 - 789221 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180473.3 NP_010450.3 1 - 1 2541 100

View Full Product Details