Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HMO1
Entrez Gene ID
851754
Gene Name
Gene Aliases
HSM2, YDR174W
Chromosome Location
Chr.IV: 812110 - 812850 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180481.1 NP_010459.1 1 - 1 450 98

View Full Product Details